missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10430-1-AP-20UL
This item is not returnable.
View return policy
Description
Cyclin-dependent kinase-5 (CDK5) is a member of the cyclin-dependent kinase family of serine/threonine kinases. Its mRNA and protein are expressed in the kidneys, testes, and ovaries. And Its activity has been detected almost exclusively in brain extracts. Similar to other Cdks, monomeric Cdk5 displays no enzymatic activity; however, Cdk5 is not activated bycyclins. Instead, Cdk5 activity requires association with one of two brain-specific regulatory subunits, called p35 and p39. These two activators regulate the spatial and temporal expression of active Cdk5, restricting its activity primarily to post-mitotic neurons.Specifications
| CDK5 | |
| Polyclonal | |
| Unconjugated | |
| CDK5 | |
| CDK5, Cell division protein kinase 5, cyclin dependent kinase 5, PSSALRE, TPKII catalytic subunit | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1020, 12568, 140908 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49615, Q00535, Q03114 | |
| Cdk5 | |
| CDK5 Fusion Protein Ag0686 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction