missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK5/p35 Recombinant Human Protein
CDK5 is a cyclin-dependent kinase that plays a critical role in neuron development.
£1434.00 - £6180.00
Specifications
Accession Number | NP_004926.1 I NP_003876 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 20% glycerol |
Molecular Weight (g/mol) | 74.5kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15428393
|
Invitrogen™
PR4026C |
100μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10210243
|
Invitrogen™
PV3981 |
1mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The CDK5 kinase is strongly activated when it complexes with neuron-specific activators, especially p35, but less strongly when it complexes with cyclins. This protein is coexpressed and copurified with p35.
Specifications
NP_004926.1 I NP_003876 | |
20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.01% Triton X-100, 2mM DTT, 20% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CDK and CHK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
74.5kDa | |
RUO - research use only | |
CDK5/p35 | |
Human | |
His Tag | |
Frozen |