missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK5 Monoclonal Antibody (DC17+DC34)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547707
This item is not returnable.
View return policy
Description
The antibody detects a 31 kDa* band corresponding to Cdk5 on SDS-PAGE immunoblots of human SYF and HeLa cells, and in mouse brain tissue. The antibody does not detect other Cdk family members, such as Cdk1 and Cdk2.
CDK5 is a member of the Cyclin-Dependent Kinase family that is most abundant in the mammalian brain. Active form of CDK5, which has also been called neuronal cdc2-like kinase, is a heterodimer of CDK5 and a 25 kDa protein which is derived proteolytically from a 35 kDa brain and neuron-specific protein and is essential for the kinase activity of CDK5. CDK5 has emerged as a crucial regulator of neuronal migration in the developing central nervous system. CDK5 phosphorylates a diverse list of substrates, implicating it in the regulation of a range of cellular processes - from adhesion and motility, to synaptic plasticity and drug addiction.
Specifications
| CDK5 | |
| Monoclonal | |
| ~0.1 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P49615, Q00535, Q03114 | |
| CDK5 | |
| Clones DC17+DC34 were generated from a recombinant full-length human Cdk5 sequence. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| DC17+DC34 | |
| Unconjugated | |
| CDK5 | |
| AW048668; Cdk5; CDK5 activator 1; CDK5-SV; CDKN5; Cell division protein kinase 5; CR6 protein kinase; Crk6; cyclin dependent kinase 5; cyclin-dependent kinase 5; cyclin-dependent-like kinase 5; LIS7; protein kinase CDK5 splicing; PSSALRE; Serine/threonine-protein kinase PSSALRE; tau protein kinase II catalytic subunit; TPKII catalytic subunit | |
| Mouse | |
| Protein A | |
| RUO | |
| 1020, 12568, 140908 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction