missing translation for 'onlineSavingsMsg'
Learn More

Cdk5 Antibody (OTI2H6), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18409394 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409394 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409394 Supplier Novus Biologicals Supplier No. NBP270396MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cdk5 Monoclonal antibody specifically detects Cdk5 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdk5
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2H6
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit
Host Species Mouse
Immunogen Full length human recombinant protein of human CDK5 (NP_004926) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Innate Immunity, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1020
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.