missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cdk5 Monoclonal antibody specifically detects Cdk5 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Cdk5 |
| Applications | Flow Cytometry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI2H6 |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Cell division protein kinase 5, cyclin-dependent kinase 5, EC 2.7.11, EC 2.7.11.22, protein kinase CDK5 splicing, PSSALRE, Serine/threonine-protein kinase PSSALRE, Tau protein kinase II catalytic subunit, TPKII catalytic subunit |
| Host Species | Mouse |
| Immunogen | Full length recombinant protein of human Cdk5 (NP_004926) produced in HEK293T cells. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?