Learn More
CDK5 Antibody Cocktail, Invitrogen™
Antibody Cocktail
Brand:  Invitrogen MA511291
Product Code. 11394413
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
MA5-11291 targets Cdk5 in IF, IP, and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-11291 immunogen is recombinant cdk5 protein.
CDK5 is a member of the Cyclin-Dependent Kinase family that is most abundant in the mammalian brain. Active form of CDK5, which has also been called neuronal cdc2-like kinase, is a heterodimer of CDK5 and a 25 kDa protein which is derived proteolytically from a 35 kDa brain and neuron-specific protein and is essential for the kinase activity of CDK5. CDK5 has emerged as a crucial regulator of neuronal migration in the developing central nervous system. CDK5 phosphorylates a diverse list of substrates, implicating it in the regulation of a range of cellular processes - from adhesion and motility, to synaptic plasticity and drug addiction.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDK5
Cocktail
0.2 mg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P49615, Q00535, Q03114
CDK5
Recombinant cdk5 protein
500 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunocytochemistry, Immunoprecipitation, Western Blot
DC17, DC34
Unconjugated
CDK5
AW048668; Cdk5; CDK5 activator 1; CDK5-SV; CDKN5; Cell division protein kinase 5; CR6 protein kinase; Crk6; cyclin dependent kinase 5; cyclin-dependent kinase 5; cyclin-dependent-like kinase 5; LIS7; protein kinase CDK5 splicing; PSSALRE; Serine/threonine-protein kinase PSSALRE; tau protein kinase II catalytic subunit; TPKII catalytic subunit
Mouse
Protein G
RUO
1020, 12568, 140908
4° C