missing translation for 'onlineSavingsMsg'
Learn More

CDK5 Activator 1 Antibody [DyLight 350], Novus Biologicals™

Product Code. 30730958 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30730958 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30730958 Supplier Novus Biologicals Supplier No. AF7250D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CDK5 Activator 1 Polyclonal antibody specifically detects CDK5 Activator 1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK5 Activator 1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CDK5 activator 1, CDK5P35, CDK5R, cyclin-dependent kinase 5 activator 1, Cyclin-dependent kinase 5 regulatory subunit 1, cyclin-dependent kinase 5, regulatory subunit 1 (p35), MGC33831, NCK5A, neuronal CDK5 activator, p23, p25, p35, p35nck5a, regulatory partner for CDK5 kinase, tau protein kinase II 23kDa subunit, TPKII regulatory subunit
Host Species Sheep
Immunogen E. coli-derived recombinant human CDK5 Activator 1, Gln100-Arg307, Accession # Q15078
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8851
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.