missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5 Activator 1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92622-0.02ml
This item is not returnable.
View return policy
Description
CDK5 Activator 1 Polyclonal antibody specifically detects CDK5 Activator 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| CDK5 Activator 1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| CDK5 activator 1, CDK5P35, CDK5R, cyclin-dependent kinase 5 activator 1, Cyclin-dependent kinase 5 regulatory subunit 1, cyclin-dependent kinase 5, regulatory subunit 1 (p35), MGC33831, NCK5A, neuronal CDK5 activator, p23, p25, p35, p35nck5a, regulatory partner for CDK5 kinase, tau protein kinase II 23kDa subunit, TPKII regulatory subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 99-307 of human CDK5R1 (NP_003876.1). AQPPPAQPPAPPASQLSGSQTGGSSSVKKAPHPAVTSAGTPKRVIVQASTSELLRCLGEFLCRRCYRLKHLSPTDPVLWLRSVDRSLLLQGWQDQGFITPANVVFLYMLCRDVISSEVGSDHELQAVLLTCLYLSYSYMGNEISYPLKPFLVESCKEAFWDRCLSVINLMSSKMLQINADPHYFTQVFSDLKNESGQEDKKRLLLGLDR | |
| 0.02 mL | |
| Cell Cycle and Replication, Signal Transduction | |
| 8851 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction