Learn More
Invitrogen™ Human CDK4/cyclin D3, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CDK4/cyclin D3 recombinant human protein (CDK4: full length, cyclin D3: full length) was expressed in insect cells. CDK4/cyclin D3 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for the subunits CKD4 and Cyclin D3 are NP_000066.1 and NP_001751.1.
Specifications
Specifications
| Accession Number | P11802, P30281 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 0.1mM PMSF, 150mM NaCl, 0.1mM EDTA, 0.25mM DTT, 25% glycerol, 10mM glutathione and no preservative; pH 7.5 |
| Gene ID (Entrez) | 1019, 896 |
| Molecular Weight (g/mol) | 60.0-58.8 kDa |
| Name | Human CDK4/cyclin D3, GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.