missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK4/CDK6 Recombinant Rabbit Monoclonal Antibody (HL2323)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556640
This item is not returnable.
View return policy
Description
CDK4/CDK6 Recombinant Monoclonal Antibody for Western Blot
CDK4 (Cyclin-Dependent Kinase 4) and CDK6 (Cyclin-Dependent Kinase 6) are crucial regulators of the cell cycle, specifically involved in the transition from the G1 phase to the S phase. These kinases form complexes with D-type cyclins (Cyclin D1, D2, and D3), which are necessary for their activation. Once activated, CDK4 and CDK6 phosphorylate the retinoblastoma protein (RB1), leading to the release of E2F transcription factors that drive the expression of genes required for DNA synthesis and cell cycle progression. Dysregulation of CDK4 and CDK6 activity is a common feature in various cancers, as it can lead to uncontrolled cell proliferation. Consequently, CDK4 and CDK6 have become important targets in cancer therapy, with inhibitors such as palbociclib, ribociclib, and abemaciclib being developed and approved for the treatment of certain types of cancer, particularly hormone receptor-positive breast cancer. These inhibitors work by blocking the kinase activity of CDK4 and CDK6, thereby halting cell cycle progression and reducing tumor growth.
Specifications
| CDK4/CDK6 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P11802, P30285, P35426, Q00534, Q64261 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1019, 1021, 114483, 12567, 12571, 94201 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| HL2323 | |
| Unconjugated | |
| CDK4 | |
| CDK4, CDK6 | |
| Recombinant fragment of human CDK4 + CDK6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction