missing translation for 'onlineSavingsMsg'
Learn More

CDK4 Antibody [DyLight 650], Novus Biologicals™

Product Code. 30735601 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735601 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735601 Supplier Novus Biologicals Supplier No. AF5254W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

CDK4 Polyclonal antibody specifically detects CDK4 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4
Host Species Goat
Immunogen E. coli-derived recombinant human CDK4, Met1-Glu303, Accession # P11802
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Protein Kinase, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1019
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.