missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK4 Antibody (CDK4/7987R), Novus Biologicals™
Rabbit Monoclonal Antibody
£242.00 - £440.00
Specifications
| Antigen | CDK4 |
|---|---|
| Clone | CDK4/7987R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18648944
|
Novus Biologicals
NBP3-20642-20ug |
20 μg |
£242.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625144
|
Novus Biologicals
NBP3-20642-100ug |
100 μg |
£440.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDK4 Monoclonal antibody specifically detects CDK4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| CDK4 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Protein Kinase, Stem Cell Markers | |
| 10mM PBS with 0.05% BSA | |
| 1019 | |
| IgG κ | |
| Protein A or G purified |
| CDK4/7987R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4 | |
| Recombinant fragment corresponding to N-terminal of human CDK4 protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title