missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK4 Antibody (6D10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42642CL1
This item is not returnable.
View return policy
Description
CDK4 Monoclonal antibody specifically detects CDK4 in Human samples. It is validated for ELISA,Western Blot
Specifications
| CDK4 | |
| Monoclonal | |
| CoraFluor 1 | |
| Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3cell division kinase 4 | |
| Recombinant human CDK4 (1-303aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Protein Kinase, Stem Cell Markers | |
| 1019 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 6D10 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction