missing translation for 'onlineSavingsMsg'
Learn More

CDK4/6-IN-2, MedChemExpress

Product Code. 16405616
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16405616

Brand: Medchem Express HY114339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CDK4/6-IN-2 is a potent CDK4 and CDK6 inhibitor extracted from patent US20180000819A1, Compound 1, has IC50s of 2.7 and 16 nM for CDK4 and CDK6, respectively.

CDK4/6-IN-2 is a potent CDK4 and CDK6 inhibitor extracted from patent US20180000819A1, Compound 1, has IC50s of 2.7 and 16 nM for CDK4 and CDK6, respectively.

Specifications

Chemical Name or Material CDK4/6-IN-2
CAS 1800506-48-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H32F2N8
Quantity 100 mg
Solubility Information DMSO : 5.2 mg/mL (10.26 mM; Need ultrasonic)
SMILES CCN1CCN(CC2=CN=C(NC3=NC=C(F)C(C4=CC(N(C(C)C)C(C)=N5)=C5C(F)=C4)=C3)N=C2)CC1
Molecular Weight (g/mol) 506.59
Formula Weight 506.59
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.