missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK2AP2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15423-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that interacts with cyclin-dependent kinase 2 associated protein 1. Pseudogenes associated with this gene are located on chromosomes 7 and 9. Alternatively spliced transcript variants have been observed for this gene.Specifications
| CDK2AP2 | |
| Polyclonal | |
| Unconjugated | |
| CDK2AP2 | |
| CDK2 associated protein 2, CDK2AP2, DOC 1 related protein, DOC 1R, DOC1R, p14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10263 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75956 | |
| CDK2AP2 | |
| CDK2AP2 Fusion Protein Ag7730 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction