missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDK20 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15713521 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15713521 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15713521 Supplier Invitrogen™ Supplier No. PA555598

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MDQYCILGRI GEGAHGIVFK AKHVETGEIV ALKKVALRRL EDGFPNQALR EIKALQEMED NQYVVQLKAV FPHG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

The protein encoded by this gene contains a kinase domain most closely related to the cyclin-dependent protein kinases. The encoded kinase may activate cyclin-dependent kinase 2 and is involved in cell growth. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK20
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDK20
Gene Accession No. Q8IZL9
Gene Alias 4932702G04Rik; AU019450; AW558027; CAK-kinase p42; Ccrk; CDCH; CDK20; CDK-activating kinase p42; CDK-related protein kinase PNQLARE; cell cycle related kinase; cell cycle-related kinase; Cell division protein kinase 20; cyclin dependent kinase 20; cyclin-dependent kinase 20; Cyclin-dependent protein kinase H; cyclin-kinase-activating kinase p42; p42; PNQALRE; PNQLARE
Gene Symbols CDK20
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDK20. Recombinant protein control fragment (Product #RP-95214).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23552
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.