missing translation for 'onlineSavingsMsg'
Learn More

CDK20 Antibody, Novus Biologicals™

Product Code. p-200049279 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423742 0.1 mL 0.1mL
18461922 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18423742 Supplier Novus Biologicals Supplier No. NBP191213

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CDK20 Polyclonal antibody specifically detects CDK20 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK20
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias CAK-kinase p42, CCRKEC 2.7.11.22, CDCHP42, CDK-activating kinase p42, cell cycle related kinase, Cell cycle-related kinase, Cell division protein kinase 20, cyclin-dependent kinase 20, Cyclin-dependent protein kinase H, Cyclin-kinase-activating kinase p42, EC 2.7.11, p42, PNQALRE
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: MDQYCILGRIGEGAHGIVFKAKHVETGEIVALKKVALRRLEDGFPNQALREIKALQEMEDNQYVVQLKAVFPHG
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23552
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.