missing translation for 'onlineSavingsMsg'
Learn More

CDK2 Antibody (AN21.2) - BSA Free, Novus Biologicals™

Product Code. 18417939 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18417939 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18417939 Supplier Novus Biologicals Supplier No. NB1002722

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CDK2 Monoclonal antibody specifically detects CDK2 in Human, Mouse, Xenopus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK2
Applications Western Blot, ELISA
Classification Monoclonal
Clone AN21.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2.5 μg/mL, ELISA 1:100 - 1:2000
Formulation PBS
Gene Alias cdc2-related protein kinase, cell devision kinase 2, Cell division protein kinase 2, cyclin-dependent kinase 2, EC 2.7.11, EC 2.7.11.22, p33 protein kinase, p33(CDK2)
Host Species Mouse
Immunogen Human recombinant CDK2
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Ovarian Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1017
Target Species Human, Mouse, Xenopus species
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.