missing translation for 'onlineSavingsMsg'
Learn More

CDK2 Antibody (6A5), Novus Biologicals™

Rabbit Monoclonal Antibody

£269.00 - £431.00

Specifications

Antigen CDK2
Clone 6A5
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043483
View Documents
Novus Biologicals
NBP3-26666-50ul
50 μL
£269.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043453
View Documents
Novus Biologicals
NBP3-26666-100ul
100 μL
£431.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CDK2 Monoclonal antibody specifically detects CDK2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications

Specifications

CDK2
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human, Mouse
cdc2-related protein kinase, cell devision kinase 2, Cell division protein kinase 2, cyclin-dependent kinase 2, EC 2.7.11, EC 2.7.11.22, p33 protein kinase, p33(CDK2)
A synthesized peptide derived from Human CDK2 [UniProt P24941]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
6A5
Western Blot, ELISA, Immunohistochemistry
Unconjugated
Rabbit
Breast Cancer, Cancer, Cell Cycle and Replication, Ovarian Carcinoma Cell Markers, Phospho Specific
PBS, pH 7.4, 150mM NaCl and 50% glycerol
1017
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.