missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDK17/cyclin Y, GST Tag Recombinant Protein

Product Code. 15970273
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15970273

Brand: Invitrogen™ A30491

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CDK17/cyclin Y recombinant human protein (CDK17: full length, cyclin Y: 2-end) was expressed in insect cells. CDK17/cyclin Y recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for CDK17/cyclin Y are NP_002586.2 and NP_659449.3.

CDK17/cyclin Y serine/threonine kinase or cyclin-dependent kinase 17 belongs to the cdc2/cdkx subfamily of kinases. CDK17/cyclin Y has similarity to a rat protein that is thought to play a role in terminally differentiated neurons. The identification of a large family of cdc2-related kinases opens the possibility of combinatorial regulation of the cell cycle together with the emerging large family of cyclins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q00537
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 10mM glutathione, 25% glycerol, 0.1mM PMSF, 150mM NaCl, 0.1mM EDTA, 0.25mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 5128
Molecular Weight (g/mol) 85.9-65.5 kDa
Name Human CDK17/cyclin Y, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 6430598J10Rik; CDK17; Cell division protein kinase 17; cyclin dependent kinase 17; cyclin-dependent kinase 17; PCTAIRE protein kinase 2; PCTAIRE2; PCTAIRE-motif protein kinase 2; PCTK2; protein kinase cdc2-related PCTAIRE-2; serine/threonine-protein kinase PCTAIRE-2
Common Name PCTAIRE2
Gene Symbol CDK17
KinaseFamily CDKs and CHKs
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence CDK17: 1-523, Cyclin Y: 2-341
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Active, Catalytic, Recombinant
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.