missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CDK16 (PCTK1)/cyclin Y, GST Tag Recombinant Protein

Product Code. 15439013
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
1mg
This item is not returnable. View return policy

Product Code. 15439013

Brand: Invitrogen™ PV6380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human CDK16 (107-end) and Cyclin Y (2-end) were co-expressed by baculovirus in Sf9 insect cells using N-terminal GST tags. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant, Fragment The gene accession number for the two subunits (CDK16[PCTK1] and Cyclin y) are NP_006192.1 and NP_659449.3.

CDK16 (PCTK1)/cyclin Y belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. CDK16 plays a role in signal transduction cascades in terminally differentiated cells, in exocytosis, and in transport of secretory cargo from the endoplasmic reticulum. CDK16 is ubiquitously expressed with the highest levels detected in the brain and testis. CDK16 also contained the gene coding for ubiquitin-activating enzyme UBE1 that previously shown to mapped to Xp11.3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q00536
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 5127
Molecular Weight (g/mol) 71.1-65.5 kDa
Name Human CDK16 (PCTK1)/cyclin Y, GST Tag
Quantity 100 μg
Gene Alias Cdk16; cdk16.L; cdk16.S; Cell division protein kinase 16; Crk5; cyclin dependent kinase 16; cyclin-dependent kinase 16; cyclin-dependent kinase 16 L homeolog; cyclin-dependent kinase 16 S homeolog; FLJ16665; PCTAIRE; PCTAIRE protein kinase 1; PCTAIRE1; PCTAIRE-1 protein kinase, alternatively spliced; PCTAIRE-motif protein kinase 1; pctgaire; pctk1; PCTK1 protein; Serine/threonine-protein kinase PCTAIRE-1; testis secretory sperm-binding protein Li 224 n
Common Name PCTAIRE1
Protein Length 108-end; 2-end
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.