Learn More
Invitrogen™ Human CDK14 (PFTK1)/cyclin Y, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human full-length CDK14 and Cyclin Y (2-end) were co-expressed by baculovirus in Sf9 insect cells using N-terminal GST tags. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Protein Form: Recombinant The gene accession number for the two subunits (CDK14[PFTK1] and Cyclin y) are NP_036527.1 and NP_659449.3.
Specifications
Specifications
| Accession Number | O94921 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 5218 |
| Molecular Weight (g/mol) | 77.0-65.5 kDa |
| Name | Human CDK14 (PFTK1)/cyclin Y, GST Tag |
| Quantity | 10 μg |
| Gene Alias | Cdk14; Cell division protein kinase 14; cyclin dependent kinase 14; cyclin-dependent kinase 14; hPFTAIRE1; Kiaa0834; mKIAA0834; PFTAIRE protein kinase 1; PFTAIRE1; Pftk1; Serine/threonine-protein kinase PFTAIRE-1 |
| Common Name | PFTK1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.