missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567681
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
CDK13/cyclin K serine/threonine kinase is dimeric protein complex. CDK13 is a member of the cyclin-dependent serine/threonine protein kinase family. Members of this family are well known for their essential roles as master switches in cell cycle control. The exact function of this protein has not yet been determined, but it may play a role in mRNA processing and may be involved in regulation of hematopoiesis. Cyclin K is a member of the transcription cyclin family. These cyclins may regulate transcription through their association with and activation of cyclin-dependent kinases (CDK) that phosphorylate the C-terminal domain of the large subunit of RNA polymerase II.
Specifications
| CDK13 | |
| Polyclonal | |
| Unconjugated | |
| Cdk13 | |
| 2310015O17Rik; CDC2L; Cdc2l5; CDC2-related protein kinase 5; CDK13; cell division cycle 2-like 5 (cholinesterase-related cell division controller); cell division cycle 2-like protein kinase 5; Cell division protein kinase 13; CHED; Cholinesterase-related cell division controller; cyclin dependent kinase 13; cyclin-dependent kinase 13; hCDK13; Kiaa1791 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 69562, 8621 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14004, Q69ZA1 | |
| Cdk13 | |
| Synthesized peptide derived from human CDK13. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction