Learn More
Invitrogen™ Human CDK13/cyclin K, GST Tag, His Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This CDK13/cyclin K recombinant human protein (CDK13: 656-1050, cyclin K 1-300) was expressed in insect cells. CDK13/cyclin K recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for CDK13 and cyclin K are NP_003709.3 and NP_001092872.1.
Specifications
Specifications
| Accession Number | O75909, Q14004 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM HEPES with 15mM glutathione, 5mM DTT, 100mM NaCl, 20% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 8621, 8812 |
| Molecular Weight (g/mol) | 73.8-63.0 kDa |
| Name | Human CDK13/cyclin K, GST Tag, His Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 100 μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.