missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDK12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114979
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CD2L7.
Plays important functions in early cell signaling. Binds the cAMP response element (CRE) (consensus: 5'-GTGACGT[AG][AG]-3'), a sequence present in many viral and cellular promoters. Activator of the NF-ELAM1/delta-A site of the E-selectin promoter. Has no intrinsic transcriptional activity, but activates transcription on formation of JUN or FOS heterodimers. Also can bind TRE promoter sequences when heterodimerized with members of the JUN family. Isoform 4/ATF-A0acts as a dominant repressor of the E-selectin/NF-ELAM1/delta-A promoter. Isoform 5/ATF-4acts as a negative regulator, inhibiting both ATF2 and ATF7 transcriptional activities. It may exert these effects by sequestrating in the cytoplasm the Thr-53 phosphorylating kinase, preventing activation.
Specifications
| CDK12 | |
| Polyclonal | |
| Unconjugated | |
| CDK12 | |
| 1810022J16Rik; AI646528; CDC2-related kinase 7; CDC2-related kinase, arginine/serine-rich; CDC2-related protein kinase 7; Cdk12; Cell division cycle 2-related protein kinase 7; cell division protein kinase 12; CRK7; CRKR; Crkrs; cyclin dependent kinase 12; cyclin-dependent kinase 12; cyclin-dependent kinase 12 isoform; D11Ertd752e; H KIAA0904; hCDK12; Kiaa0904; Pksc; Protein kinase for splicing component | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 192350, 51755 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q3MJK5, Q9NYV4 | |
| CDK12 | |
| A synthesized peptide derived from human CDK12(Accession Q9NYV4), corresponding to amino acid residues L1213-E1263. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction