missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDK11B Polyclonal Antibody

Product Code. 15517551
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15517551 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15517551 Supplier Invitrogen™ Supplier No. PA536154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 110 and 92 kDa. Purity is >95% by SDS-PAGE.

The PITSLRE beta1 protein, a distantly related member of the Cdk family of protein kinases, induces apoptosis after low levels of ectopic expression. Apoptosis, or programmed cell death, is similarly induced by ectopic expression of an amino terminal deletion mutant retaining the catalytic and carboxyterminal domains of PITSLRE beta1, but not by other mutants lacking Histone H1 kinase activity or by other Cdk family members. The terminology for the ten isoforms of the PITSLRE subfamily of proteins is based on the conserved PSTAIRE box region of Cdc2 p34. Depending on which of the PITSLRE genes produce the protein, the cDNA and protein are designated alpha, beta or gamma (i.e., PITSLRE A gene, alpha; PITSLRE B gene, beta and PITSLRE C gene, gamma). Some of the isoforms such as PITSLRE alpha1 (T cells) and PITSLRE beta1 (B cells and brain), are expressed in specific cell types, while others are expressed ubiquitously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK11B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene CDK11B
Gene Accession No. P21127, P24788, P46892
Gene Alias AA989746; Cdc11b; CDC2L1; Cdc2l2; CDC2L3; CDC-related protein kinase p58; CDK11; CDK11 p110; CDK11 p46; CDK11 p58; CDK11A; Cdk11b; CDK11-p110; CDK11-p46; CDK11-p58; cell division cycle 2 homolog-like 1; cell division cycle 2 homolog-like 2; cell division cycle 2-like 1; cell division cycle 2-like 1 (PITSLRE proteins); cell division cycle 2-like 2; cell division cycle 2-like protein kinase 1; Cell division protein kinase 11; cell division protein kinase 11B; CLK-1; cyclin dependent kinase 11B; cyclin-dependent kinase 11; Cyclin-dependent kinase 11B; galactosyltransferase-associated protein kinase p58/GTA; p58; p58 CLK-1; p58CDC2L1; p58CLK-1; PITSLRE proteins; PITSLRE serine/threonine-protein kinase CDC2L1; PITSLREA; PK58
Gene Symbols CDK11B
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of human PITSLRE.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12537, 252879, 984
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.