missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55913
This item is not returnable.
View return policy
Description
CDK10 Polyclonal specifically detects CDK10 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| CDK10 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| Cell division protein kinase 10, cyclin-dependent kinase (CDC2-like) 10, cyclin-dependent kinase 10, cyclin-dependent kinase related protein, EC 2.7.11, EC 2.7.11.22, PISSLRECDC2-related protein kinase, serine/threonine protein kinase PISSLRE, Serine/threonine-protein kinase PISSLRE | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| CDK10 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:YFKEKPLPCEPELMPTFPHHRNKRAAP | |
| 100 μL | |
| Cell Cycle and Replication | |
| 8558 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction