missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38008-20ul
This item is not returnable.
View return policy
Description
CDK10 Polyclonal antibody specifically detects CDK10 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| CDK10 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Cell division protein kinase 10, cyclin-dependent kinase (CDC2-like) 10, cyclin-dependent kinase 10, cyclin-dependent kinase related protein, EC 2.7.11, EC 2.7.11.22, PISSLRECDC2-related protein kinase, serine/threonine protein kinase PISSLRE, Serine/threonine-protein kinase PISSLRE | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human CDK10 (NP_001092003.2).,, Sequence:, MDKEKDGIPISSLREITLLLRLRHPNIVELKEVVVGNHLESIFLVMGYCEQDLASLLENMPTPFSEAQVKCIVLQVLRGLQYLHRNFIIHRDLKVSNLLMTDKGCVKTADFGLARAYGVPVKPMTPKVVTLWYRAPELLLGTTTQTTSIDMWAVGCILAELLAHRPLLPGTSEIHQIDLIVQLLGTPSENIWPGFSKLPLVGQYSLRKQPYNNLKHKFPWLSEAGLRLLHFLFMATAGDCLESSYFKEKPLPCEPELMPTFPHHRNKRAAPATSEGQSKRCKP | |
| 20 μL | |
| Cell Cycle and Replication | |
| 8558 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction