missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561940
This item is not returnable.
View return policy
Description
Immunogen sequence: CLINDFKDVT HTCPSCKAYI YTYKLLC Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
The p53 tumor-suppressor gene integrates numerous signals that control cell life and death; loss of its functions contributes to the development of most cancers. CDIP is a novel pro-apoptotic target gene whose inhibition abrogates p53-mediated apoptotic responses. Overexpression of CDIP induced apoptosis in transfected cells while siRNA suppression of caspase-8 mRNA blocked this CDIP-induced apoptosis, indicating that the CDIP-dependent apoptosis pathway proceeds through extrinsic cell death pathway. CDIP may thus represent a novel target for drug design to maximize p53 response and sensitize tumor cells to cancer therapy. Multiple isoforms of CDIP are known to exist.
Specifications
| CDIP1 | |
| Polyclonal | |
| Unconjugated | |
| CDIP1 | |
| 2700048O17Rik; 5730403B10Rik; AU018521; AW742692; C16orf5; CDIP; CDIP1; cell death inducing protein; cell death inducing Trp53 target 1; cell death involved p53-target; cell death-inducing p53 target 1; cell death-inducing p53-target protein 1; Cell death-inducing protein; I1; Lipopolysaccharide-induced tumor necrosis factor-alpha-like protein; LITAFL; LITAF-like protein; Transmembrane protein I1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29965 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H305 | |
| CDIP1 | |
| Recombinant protein corresponding to Human CDIP1. Recombinant protein control fragment (Product #RP-101138). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction