missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDHR3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA578657

Product Code. 16317354

  • £463.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: transfected CDHR3 in HEK293FT Predicted Reactivity: Mouse (80%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cadherins are calcium dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell type.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CDHR3
Polyclonal
Unconjugated
CDHR3
Cadherin; cadherin related family member 3; cadherin-like protein 28; Cadherin-related family member 3; CDH28; CDHR3; FLJ23834; FLJ43271; MGC133292; MGC133293
Rabbit
Antigen affinity chromatography
RUO
222256
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q6ZTQ4
CDHR3
Recombinant protein encompassing a sequence within the Extracellular domain of human FLJ23834. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.