missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDH8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581597
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Rat Cadherin-8/CDH8.
CDH8 is a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. The extracellular domain consists of 5 subdomains, each containing a cadherin motif, and appears to determine the specificity of the protein's homophilic cell adhesion activity. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is expressed in brain and is putatively involved in synaptic adhesion, axon outgrowth and guidance.
Specifications
| CDH8 | |
| Polyclonal | |
| Unconjugated | |
| CDH8 | |
| AI851472; cad8; CADH8; Cadherin; cadherin 8; cadherin 8, type 2; Cadherin8; cadherin-8; Cdh8; Nbla04261 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 84408 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| O54800 | |
| CDH8 | |
| Recombinant Rat Cadherin-8/CDH8 Protein (Met1-Met621). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction