missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDH26, Mouse anti-Human, Clone: 6C10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00060437-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: MKPLIWTWSDVEGQRPALLICTAAAGPTQGVKDLEEVPPSAASQSAQARCALGSWGYGKPFEPRSVKNIHSTPAYPDATMHRQLLAPVEGRMAETLNQKLHVANVLEDDPGYLPHVYSEEGECGGAPSLSSLASLEQELQPDLLDSLGSKATPFEEIYSESGVPSSpecifications
| CDH26 | |
| Monoclonal | |
| Unconjugated | |
| cadherin-like 26 | |
| VR20 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 6C10 | |
| In 1x PBS, pH 7.4 | |
| NM_021810.3 | |
| CDH26 | |
| CDH26 (NP_068582.2, 1 a.a. ∼ 165 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 60437 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction