missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDH24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584685
This item is not returnable.
View return policy
Description
Immunogen sequence: TVLATELGWS WGPERGWVPL LVAEWSAPAA PPQRSPVGSA VGIPQDSSAQ AS Highest antigen sequence indentity to the following orthologs: Rat - 31%, Mouse - 31%.
Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types. Cadherin-24 mediate strong cell-cell adhesion.
Specifications
| CDH24 | |
| Polyclonal | |
| Unconjugated | |
| CDH24 | |
| 1700040A22Rik; Cadherin; cadherin 24; cadherin 24, type 2; cadherin-24; cadherin-like 24; CDH11L; CDH24; ENSMUSG00000022188; EY-cadherin; UNQ2834/PRO34009 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64403 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86UP0 | |
| CDH24 | |
| Recombinant protein corresponding to Human CDH24. Recombinant protein control fragment (Product #RP-106510). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction