missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDH23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13496-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of the cadherin superfamily, whose genes encode calcium dependent cell-cell adhesion glycoproteins. The encoded protein is thought to be involved in stereocilia organization and hair bundle formation. The gene is located in a region containing the human deafness loci DFNB12 and USH1D. Usher syndrome 1D and nonsyndromic autosomal recessive deafness DFNB12 are caused by allelic mutations of this cadherin-like gene. Alternative splice variants encoding different isoforms have been described.Specifications
| CDH23 | |
| Polyclonal | |
| Unconjugated | |
| CDH23 | |
| CAD23, Cadherin 23, cadherin like 23, CDH23, DFNB12, KIAA1774, KIAA1812, Otocadherin, USH1D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114102, 22295, 64072 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P58365, Q99PF4, Q9H251 | |
| CDH23 | |
| CDH23 Fusion Protein Ag4322 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction