missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDH22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101292
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CDH22.
This gene is a member of the cadherin superfamily. The gene product is composed of five cadherin repeat domains and a cytoplasmic tail similar to the highly conserved cytoplasmic region of classical cadherins. Expressed predominantly in the brain, this putative calcium-dependent cell adhesion protein may play an important role in morphogenesis and tissue formation in neural and non-neural cells during development and maintenance of the brain and neuroendocrine organs. [provided by RefSeq, Jul 2008].
Specifications
| CDH22 | |
| Polyclonal | |
| Unconjugated | |
| CDH22 | |
| C20orf25; Cadherin; cadherin 22; cadherin 22, type 2; cadherin-22; cadherin-like 22; CDH22; dJ998H6.1; ortholog of rat PB-cadherin; PB-cadherin; Pituitary and brain cadherin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 104010, 29182, 64405 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q63315, Q9UJ99, Q9WTP5 | |
| CDH22 | |
| A synthesized peptide derived from human CDH22(Accession Q9UJ99), corresponding to amino acid residues E114-S164. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction