Learn More
Invitrogen™ CDH18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553162
Description
Immunogen sequence: DNPPRFPQKH YQLYVPESAQ VGSAVGKIKA NDADTGSNAD MTYSIINGDG MGIFSISTDK ETREGILSLK KPLNYEKKKS YTLNIEGANT HLDFRFSHLG PFKDATMLKI IVGDVDEPPL FSMPSYLMEV YENAKIGTVV GTVLAQDPD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.
This gene encodes a type II classical cadherin from the cadherin superfamily of integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is expressed specifically in the central nervous system and is putatively involved in synaptic adhesion, axon outgrowth and guidance. Cadherins are calcium dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types.
Specifications
| CDH18 | |
| Polyclonal | |
| Unconjugated | |
| CDH18 | |
| B230220E17Rik; Cadherin; Cadherin 14; cadherin 18; cadherin 18, type 2; Cadherin14; cadherin-14; Cadherin18; Cadherin-18; cadherin-like 24; CDH14; Cdh14l; CDH18; CDH24; ey-cadherin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1016 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13634 | |
| CDH18 | |
| Recombinant protein corresponding to Human CDH18. Recombinant protein control fragment (Product #RP-90592). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.