missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDH11, Mouse anti-Human, Clone: 2C67, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00001009-M25.100ug
This item is not returnable.
View return policy
Description
Sequence: GWVWNQFFVIEEYTGPDPVLVGRLHSDIDSGDGNIKYILSGEGAGTIFVIDDKSGNIHATKTLDREERAQYTLMAQAVDRDTNRPLEPPSEFIVKVQDINDNPPEFLHETYHANVPERSNVGTSVIQVTASDADDPTYGNSAKLVYSILEGQPYFSVEAQTGIIRTALPNMDREAKEEYHVVIQAKDMGGHMGGLSGTTKVMITLTDVNDNPPKFPQSVYQMSVSEAAVPGEEVGRVKAKDPDIGENGLVTYNIVDGDGMESFEITTDYETQEGVIKLKKPVDFETKRAYSLKVEAANVHIDPKFISNGPFKDTVTVKIAVEDADEPPMFLAPSYIHEVQENAAAGTVVGRVHAKDPDAANSPIRYSIDRHTDLDRFFTINPEDGFIKTTKPLDREETAWLNITVFAAEIHNRHQEAKVPVAIRVLDVNDNAPKFAAPYEGFICESDQTKPLSNQPIVTISADDKDDTANGPRFIFSLPPEIIHNPNFTVRDNRDNTAGVYARRGGFSRQKQDLYLLPIVISDGGIPPMSSTNTLTIKVCGCDVNGALLSCNAEAYILNSpecifications
| CDH11 | |
| Monoclonal | |
| Unconjugated | |
| cadherin 11, type 2, OB-cadherin (osteoblast) | |
| CAD11/CDHOB/OB/OSF-4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2C67 | |
| In 1x PBS, pH 7.4 | |
| BC013609.1 | |
| CDH11 | |
| CDH11 (P55287, 54 a.a. ∼ 612 a.a) partial recombinant protein with Fc tag at C terminal. MW of the Fc tag alone is 50 KDa. | |
| RUO | |
| 1009 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction