missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDH10 Recombinant Rabbit Monoclonal Antibody (K01_2L62)
Description
CDH10 Recombinant Monoclonal Antibody for Western Blot
CDH10 is a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. The extracellular domain consists of 5 subdomains, each containing a cadherin motif, and appears to determine the specificity of the protein's homophilic cell adhesion activity. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is predominantly expressed in brain and is putatively involved in synaptic adhesions, axon outgrowth and guidance.
Specifications
Specifications
| Antigen | CDH10 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2L62 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | Cdh10 |
| Gene Accession No. | P70408, Q9Y6N8 |
| Gene Alias | A830016G23Rik; C030003B10Rik; C030011H18Rik; Cadherin; cadherin 10; cadherin 10 type 2; cadherin 10, type 2 (T2-cadherin); cadherin-10; Cdh10; T2-cadherin |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?