missing translation for 'onlineSavingsMsg'
Learn More

CDD3505, MedChemExpress

Product Code. 16464486
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464486 100 mg 100mg
16455947 5 mg 5mg
16433218 10 mg 10mg
16429766 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464486 Supplier Medchem Express Supplier No. HY100901
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CDD3505 is used for elevating high density lipoprotein cholesterol (HDL) by inducing hepatic cytochrome P450IIIA (CYP3A) activity.

CDD3505 is used for elevating high density lipoprotein cholesterol (HDL) by inducing hepatic cytochrome P450IIIA (CYP3A) activity.

Specifications

Chemical Name or Material CDD3505
CAS 173865-33-3
Color White
Purity Grade Notes Research
Molecular Formula C22H17N3O2
Quantity 100 mg
Solubility Information DMSO : 14.29 mg/mL (40.21 mM; Need ultrasonic)
SMILES O=[N+]([O-])C1=CN(C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)C=N1
Molecular Weight (g/mol) 355.39
Formula Weight 355.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.