missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDCP1 Monoclonal Antibody (OTI2B8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525357
This item is not returnable.
View return policy
Description
CDCP1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
CDCP1 is a transmembrane protein containing three extracellular CUB domains. It is found to be overexpressed in colon and lung cancers. Its expression level is correlated with the metastatic ability of carcinoma cells. CDCP1 is located on the cell surface. It has been shown to be tyrosine phosphorylated in a cancer cell line. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| CDCP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H5V8 | |
| Cdcp1 | |
| Full length human recombinant protein of CDCP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI2B8 | |
| Unconjugated | |
| Cdcp1 | |
| 9030022E12Rik; AA409659; CD318; CDCP1; CUB domain containing protein 1; CUB domain-containing protein 1; E030027H19Rik; gp140; Membrane glycoprotein gp140; RGD1305578; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; transmembrane and associated with src kinases; TRASK; UNQ2486/PRO5773 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64866 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction