missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDCA8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102318
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CDCA8.
CDCA8 is a component of a chromosomal passenger complex (CPC) required for stability of the bipolar mitotic spindle. The chromosomal passenger complex, which includes Survivin, CDCA8, INCENP and Aurora-B, is known to play crucial roles during mitosis and cell division. It was found that CDCA8 interacting with Aurora-B, INCENP and Survivin, increases during G2/M phase and then reduces after exit from M phase. CDCA8 is cell cycle regulated, down-regulated in response to p53/Rb-signaling, and up-regulated in many types of cancerous tissues. In Drosophila cells, inactivation of CDCA8 results in polyploidy, delayed mitosis and abnormal tissue development, indicating its critical role for cell proliferation. Recent studies show that CDCA8 is essential for cell proliferation during early embryonic development, and its early embryonic lethality cannot be rescued by the loss of p53. Its aberrant expression is linked to a poor prognosis for gastric cancer.
Specifications
| CDCA8 | |
| Polyclonal | |
| Unconjugated | |
| CDCA8 | |
| 4831429J16Rik; AU044747; BOR; Borealin; Cdca8; cell division cycle associated 8; cell division cycle-associated protein 8; D4Ertd421e; Dasra B; DasraB; dasra-B; embryonic stem cell-related; hDasra-B; MESrg; MESRGP; PESCRG3; Pluripotent embryonic stem cell-related gene 3 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 500545, 52276, 55143 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q53HL2, Q6AXW0, Q8BHX3 | |
| CDCA8 | |
| A synthesized peptide derived from human CDCA8(Accession Q53HL2), corresponding to amino acid residues M1-V32. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction