missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDCA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545892
This item is not returnable.
View return policy
Description
Peptide sequence: CRTRSQCRHS GPLRVAMKFP ARSTRGATNK KAESRQPSEN SVTDSNSDSE Sequence homology: Human: 100%.
This gene was identified as a c-Myc responsive gene, and behaves as a direct c-Myc target gene. Overexpression of this gene is found to enhance the transformation of lymphoblastoid cells, and it complements a transformation-defective Myc Box II mutant, suggesting its involvement in c-Myc-mediated cell transformation. Two alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| CDCA7 | |
| Polyclonal | |
| Unconjugated | |
| CDCA7 | |
| 2310021G01Rik; AU044109; CDCA7; cell division cycle associated 7; cell division cycle-associated protein 7; c-Myc target JPO1; ICF3; JPO1; Protein JPO1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83879 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BWT1 | |
| CDCA7 | |
| Synthetic peptide directed towards the N-terminal of human CDCA7 (aa 58-107). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction