missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55144
This item is not returnable.
View return policy
Description
CDCA5 Polyclonal specifically detects CDCA5 in Human samples. It is validated for Western Blot.
Specifications
| CDCA5 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| cell division cycle associated 5, Cell division cycle-associated protein 5, MGC16386, p35, SORORIN | |
| Rabbit | |
| 28 kDa | |
| 100 μL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 113130 | |
| Human | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96FF9 | |
| CDCA5 | |
| Synthetic peptides corresponding to CDCA5(cell division cycle associated 5) The peptide sequence was selected from the middle region of CDCA5. Peptide sequence ATPTSTPVPNPEAESSSKEGELDARDLEMSKKVRRSYSRLETLGSASTST. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction