missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11625-1-AP-150UL
This item is not returnable.
View return policy
Description
CDCA4 encodes a protein that belongs to the E2F family of transcription factors. This protein regulates E2F-dependent transcriptional activation and cell proliferation, mainly through the E2F/retinoblastoma protein pathway. It also functions in the regulation of JUN oncogene expression. This protein shows distinctive nuclear-mitotic apparatus distribution, it is involved in spindle organization from prometaphase, and may also play a role as a midzone factor involved in chromosome segregation or cytokinesis. Two alternatively spliced transcript variants encoding the same protein have been noted for this gene. Two pseudogenes have also been identified on chromosome 1.Specifications
| CDCA4 | |
| Polyclonal | |
| Unconjugated | |
| CDCA4 | |
| CDCA4, HEPP, SEI 3/HEPP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55038, 71963 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BXL8, Q9CWM2 | |
| CDCA4 | |
| CDCA4 Fusion Protein Ag2214 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction