Learn More
Invitrogen™ CDCA3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA513564
Product Code. 10893401
  • £479.00 / 400µL
Description
CDCA3 Polyclonal Antibody for Western Blot, IHC (P), Flow
F-box-like protein which is required for entry into mitosis. Acts by participating in E3 ligase complexes that mediate the ubiquitination and degradation of WEE1 kinase at G2/M phase. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDCA3
Polyclonal
Unconjugated
CDCA3
2410005A12Rik; C8; CDCA 3; CDCA3; cell division cycle associated 3; cell division cycle-associated protein 3; gene rich cluster, C8; gene-rich cluster protein C8; GRCC 8; GRCC8; MGC2577; TOME 1; Tome1; TOME-1; trigger of mitotic entry 1; trigger of mitotic entry protein 1
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
14793, 83461
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q99618, Q99M54
CDCA3
KLH conjugated synthetic peptide between 41-69 amino acids from the N-terminal region of human CDCA3.
400 μL
Primary
Human, Mouse