missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc73 Recombinant Rabbit Monoclonal Antibody (23GB3450), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Cdc73 |
|---|---|
| Clone | 23GB3450 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088946
![]() |
Invitrogen™
MA553424 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089272
|
Invitrogen™
MA553425 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cdc73 Recombinant Monoclonal antibody specifically detects Cdc73 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Cdc73 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q4V8C8, Q6P1J9, Q8JZM7 | |
| 214498, 304832, 79577 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3450 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 8430414L16Rik; BC027756; C130030P16Rik; C1orf28; C81219; Cdc73; cell division cycle 73; cell division cycle 73 Paf1/RNA polymerase II complex component-like protein; cell division cycle 73, Paf1/RNA polymerase II complex component; cell division cycle 73, Paf1/RNA polymerase II complex component, homolog; cell division cycle protein 73 homolog; FIHP; HPTJT; HRPT1; HRPT2; hyperparathyroidism 2 (with jaw tumor); hyperparathyroidism 2 homolog; hyperparathyroidism 2 protein; Hyperparathyroidism 2 protein homolog; HYX; Paf1/RNA polymerase II complex component; parafibromin; Vcell division cycle 73, Paf1/RNA polymerase II complex component, homolog | |
| A synthetic peptide of human HRPT2/CDC73 (350-400AA). | |
| Primary | |
| -20°C | |
| CDC73 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title