Learn More
Invitrogen™ Cdc73 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529114
Description
Recommended positive controls: 293T, A431, H1299, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (90%), Xenopus laevis (91%), Dog (100%), Chicken (97%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cdc73 is a tumor suppressor probably involved in transcriptional and post-transcriptional control pathways. Cdc73 is part of the PAF1 complex, a five-subunit protein complex composed of Paf1, Cdc73, Leo1, Rtf1 and Ctr9. PAF1 was first purified from yeast in association with RNA polymerase II and is believed to function in transcription elongation. The yeast PAF (yPAF) complex interacts with RNA polymerase II and coordinates the setting of histone marks associated with active transcription. In addition to coordinating events during transcription (initiation, promoter clearance, and elongation), hPAF also coordinates events in RNA quality control. Cdc73 may also be involved in cell cycle progression through the regulation of cyclin D1/PRAD1 expression.
Specifications
| Cdc73 | |
| Polyclonal | |
| Unconjugated | |
| CDC73 | |
| 8430414L16Rik; BC027756; C130030P16Rik; C1orf28; C81219; Cdc73; cell division cycle 73; cell division cycle 73 Paf1/RNA polymerase II complex component-like protein; cell division cycle 73, Paf1/RNA polymerase II complex component; cell division cycle 73, Paf1/RNA polymerase II complex component, homolog; cell division cycle protein 73 homolog; FIHP; HPTJT; HRPT1; HRPT2; hyperparathyroidism 2 (with jaw tumor); hyperparathyroidism 2 homolog; hyperparathyroidism 2 protein; Hyperparathyroidism 2 protein homolog; HYX; Paf1/RNA polymerase II complex component; parafibromin; Vcell division cycle 73, Paf1/RNA polymerase II complex component, homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79577 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.89 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q6P1J9 | |
| CDC73 | |
| Recombinant protein encompassing a sequence within the center region of human CDC73. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.