missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC73/HRPT2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-184IR
This item is not returnable.
View return policy
Description
CDC73/HRPT2 Polyclonal antibody specifically detects CDC73/HRPT2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CDC73/HRPT2 | |
| Polyclonal | |
| Western Blot | |
| C1orf28hyperparathyroidism 2 (with jaw tumor), cell division cycle 73, Paf1/RNA polymerase II complex component, homolog (S.cerevisiae), Cell division cycle protein 73 homolog, chromosome 1 open reading frame 28, HPTJT, HRPT2FLJ23316, Hyperparathyroidism 2 protein, HYX, parafibromin | |
| The immunogen recognized by this antibody maps to a region between residues 250 and 300 of human Parafibromin using the numbering given in TrEMBL entry Q6P1J9 (GeneID 79577). | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 79577 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction