missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CDC7 Recombinant Rabbit Monoclonal Antibody (K01_1P34)

Product Code. 30827720
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827720 100 μL
30825164 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827720 Supplier Invitrogen™ Supplier No. MA563573
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

CDC7 Recombinant Monoclonal Antibody for Western Blot

CDC7 is a cell division cycle 7 homolog protein that is critical for the G1/S transition and is also essential for initiation of DNA replication as cell division occurs. CDC7 is expressed in many normal tissues, but the overexpression of CDC7 may be associated with neoplastic transformation for some tumors and transformed cell lines. CDC7/DBF4 kinase promotes S phase by alleviating an inhibitory activity in Mcm4 that evolved to integrate several protein kinase. CDC7 encodes a cell division cycle protein with kinase activity that is critical for the G1/S transition. The yeast homolog is also essential for initiation of DNA replication as cell division occurs. Overexpression of this gene product may be associated with neoplastic transformation for some tumors. Multiple alternatively spliced transcript variants that encode the same protein have been detected.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDC7
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1P34
Concentration 0.70 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene CDC7
Gene Accession No. O00311
Gene Alias AI597260; Cdc7; CDC7 (cell division cycle 7, S. cerevisiae, homolog)-like 1; Cdc7 Kinase; Cdc7l1; CDC7-like 1; CDC7-related kinase; cell division cycle 7; cell division cycle 7 (S. cerevisiae); cell division cycle 7 homolog; cell division cycle 7-like 1; cell division cycle 7-like protein 1; cell division cycle 7-related protein kinase; HsCDC7; Hsk1; huCDC7; MGC117361; MGC126237; MGC126238; MGC91067; muCdc7
Gene Symbols CDC7
Host Species Rabbit
Immunogen Recombinant protein of human CDC7.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100689341, 8317
Target Species Hamster, Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.