missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cdc7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA124588
This item is not returnable.
View return policy
Description
PA1-24588 detects CDC7-Related Protein Kinase (CDC7L1) in Human samples.
CDC7 is a cell division cycle 7 homolog protein that is critical for the G1/S transition and is also essential for initiation of DNA replication as cell division occurs. CDC7 is expressed in many normal tissues, but the overexpression of CDC7 may be associated with neoplastic transformation for some tumors and transformed cell lines. CDC7/DBF4 kinase promotes S phase by alleviating an inhibitory activity in Mcm4 that evolved to integrate several protein kinase. CDC7 encodes a cell division cycle protein with kinase activity that is critical for the G1/S transition. The yeast homolog is also essential for initiation of DNA replication as cell division occurs. Overexpression of this gene product may be associated with neoplastic transformation for some tumors. Multiple alternatively spliced transcript variants that encode the same protein have been detected.
Specifications
| Cdc7 | |
| Polyclonal | |
| Unconjugated | |
| CDC7 | |
| AI597260; Cdc7; CDC7 (cell division cycle 7, S. cerevisiae, homolog)-like 1; Cdc7 Kinase; Cdc7l1; CDC7-like 1; CDC7-related kinase; cell division cycle 7; cell division cycle 7 (S. cerevisiae); cell division cycle 7 homolog; cell division cycle 7-like 1; cell division cycle 7-like protein 1; cell division cycle 7-related protein kinase; HsCDC7; Hsk1; huCDC7; MGC117361; MGC126237; MGC126238; MGC91067; muCdc7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100060468, 100343407, 479955, 516094, 8317 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O00311 | |
| CDC7 | |
| Synthetic 19 amino acid peptide from kinase domain of human CDC7. | |
| 25 μg | |
| Primary | |
| Human, Canine, Rabbit, Bovine, Horse, Bat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto